Assay Details
| Gene Symbol: B4galt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246694.2 | NP_001233623.1 | 5 - 6 | 965 | 76 | |
| XM_007643277.1 | XP_007641467.1 | 6 - 7 | 1845 | 76 | ||
| GenBank mRNA | AY117536.1 | - | 5 - 6 | 968 | 76 | |
| JI896241.1 | - | 4 - 5 | 445 | 76 | ||
| JP059857.1 | - | 4 - 5 | 445 | 76 | ||
More Information
Set Membership: |
|

B4galt2
Genomic Map