Assay Details
| Gene Symbol: Adi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285925.1 | NP_001272854.1 | 3 - 4 | 419 | 58 | |
| GenBank mRNA | JI891561.1 | - | 2 - 3 | 221 | 58 | |
| JP039169.1 | - | 2 - 3 | 221 | 58 | ||
More Information
Set Membership: |
|

Adi1
Genomic Map