Assay Details
| Gene Symbol: Gnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246701.1 | NP_001233630.1 | 8 - 9 | 784 | 65 | |
| XM_007649854.1 | XP_007648044.1 | 8 - 9 | 797 | 65 | ||
| GenBank mRNA | AY392427.1 | - | 8 - 9 | 784 | 65 | |
| JI891102.1 | - | 8 - 9 | 827 | 65 | ||
| JP055946.1 | - | 8 - 9 | 827 | 65 | ||
More Information
Set Membership: |
|

Gnb1
Genomic Map