Assay Details
| Gene Symbol: Prdx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246780.2 | NP_001233709.1 | 5 - 6 | 534 | 68 | |
| GenBank mRNA | AF532110.1 | - | 5 - 6 | 534 | 68 | |
| JI877296.1 | - | 5 - 6 | 646 | 68 | ||
| JP044317.1 | - | 5 - 6 | 646 | 68 | ||
More Information
Set Membership: |
|

Prdx2
Genomic Map