Assay Details
| Gene Symbol: Selt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256851.1 | NP_001243780.1 | 5 - 6 | 699 | 62 | |
| GenBank mRNA | JI864277.1 | - | 5 - 6 | 671 | 62 | |
| JP065339.1 | - | 5 - 6 | 671 | 62 | ||
More Information
Set Membership: |
|

Selt
Genomic Map