Assay Details
| Gene Symbol: LOC100759157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001292060.1 | NP_001278989.1 | 22 - 23 | 2490 | 108 | |
| XM_007647583.1 | XP_007645773.1 | 22 - 23 | 2493 | 108 | ||
More Information
Set Membership: |
|

LOC100759157
Genomic Map