Assay Details
| Gene Symbol: LOC100759157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001292060.1 | NP_001278989.1 | 6 - 7 | 509 | 71 | |
| XM_007647583.1 | XP_007645773.1 | 6 - 7 | 509 | 71 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100759157| Gene Symbol: LOC100759157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001292060.1 | NP_001278989.1 | 6 - 7 | 509 | 71 | |
| XM_007647583.1 | XP_007645773.1 | 6 - 7 | 509 | 71 | ||