Assay Details
| Gene Symbol: Abcb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243988.1 | NP_001230917.1 | 16 - 17 | 2150 | 70 | |
| XM_007646772.1 | XP_007644962.1 | 16 - 17 | 2120 | 70 | ||
| XM_007646773.1 | XP_007644963.1 | 13 - 14 | 1798 | 70 | ||
| GenBank mRNA | JI893849.1 | - | 1716 | 70 | ||
| JP062803.1 | - | 10 - 11 | 1437 | 70 | ||
| M60040.1 | - | 16 - 17 | 2150 | 70 | ||
More Information
Set Membership: |
|

Abcb1a
Genomic Map