Assay Details
| Gene Symbol: Ndufb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246789.1 | NP_001233718.1 | 1 - 2 | 244 | 68 | |
| GenBank mRNA | AY649405.1 | - | 1 - 2 | 244 | 68 | |
| JI883463.1 | - | 1 - 2 | 266 | 68 | ||
| JP054321.1 | - | 1 - 2 | 265 | 68 | ||
More Information
Set Membership: |
|

Ndufb11
Genomic Map