Assay Details
| Gene Symbol: Atf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246812.1 | NP_001233741.1 | 1 - 2 | 221 | 64 | |
| XM_007654285.1 | XP_007652475.1 | 2 - 3 | 679 | 64 | ||
| GenBank mRNA | AB300203.1 | - | 1 - 2 | 221 | 64 | |
| JI879957.1 | - | 2 - 3 | 497 | 64 | ||
More Information
Set Membership: |
|

Atf4
Genomic Map