Assay Details
| Gene Symbol: Rps13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244533.1 | NP_001231462.1 | 4 | 226 | 90 | |
| GenBank mRNA | JI886075.1 | - | 4 | 297 | 90 | |
| JP044083.1 | - | 4 | 297 | 90 | ||
More Information
Set Membership: |
|

Rps13
Genomic Map