Assay Details
| Gene Symbol: Ubl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499750.2 | XP_003499798.1 | 3 - 4 | 271 | 62 | |
| XM_007640509.1 | XP_007638699.1 | 2 - 3 | 154 | 62 | ||
| XM_007640510.1 | XP_007638700.1 | 2 - 3 | 514 | 62 | ||
| GenBank mRNA | JI890406.1 | - | 1 - 2 | 87 | 62 | |
More Information
Set Membership: |
|

Ubl5
Genomic Map