Assay Details
| Gene Symbol: Chmp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503915.2 | XP_003503963.1 | 5 - 6 | 660 | 60 | |
| GenBank mRNA | JI883004.1 | - | 5 - 6 | 617 | 60 | |
More Information
Set Membership: |
|

Chmp3
Genomic Map