Assay Details
| Gene Symbol: Ppp6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649140.1 | XP_007647330.1 | 2 - 3 | 132 | 71 | |
| GenBank mRNA | JI877870.1 | - | 2 - 3 | 174 | 71 | |
More Information
Set Membership: |
|

Ppp6c
Genomic Map