Assay Details
| Gene Symbol: Lrpap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496604.2 | XP_003496652.1 | 1 - 2 | 290 | 65 | |
| XM_007651602.1 | XP_007649792.1 | 1 - 2 | 290 | 65 | ||
| GenBank mRNA | JI884965.1 | - | 1 - 2 | 264 | 65 | |
| JP063167.1 | - | 1 - 2 | 264 | 65 | ||
More Information
Set Membership: |
|

Lrpap1
Genomic Map