Assay Details
| Gene Symbol: Mrps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515101.2 | XP_003515149.2 | 3 - 4 | 274 | 62 | |
| GenBank mRNA | JI888489.1 | - | 2 - 3 | 202 | 62 | |
More Information
Set Membership: |
|

Mrps2
Genomic Map