Assay Details
| Gene Symbol: Atp5g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498323.2 | XP_003498371.1 | 1 - 2 | 177 | 80 | |
| GenBank mRNA | JI896039.1 | - | 1 - 2 | 177 | 80 | |
| JP060363.1 | - | 1 - 2 | 177 | 80 | ||
More Information
Set Membership: |
|

Atp5g3
Genomic Map