Assay Details
| Gene Symbol: Tiparp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498406.2 | XP_003498454.2 | 2 - 3 | 1178 | 75 | |
| GenBank mRNA | JI870725.1 | - | 1 - 2 | 165 | 75 | |
| JP047815.1 | - | 1 - 2 | 165 | 75 | ||
More Information
Set Membership: |
|

Tiparp
Genomic Map