Assay Details
| Gene Symbol: LOC100773059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509677.1 | XP_003509725.1 | 2 - 3 | 165 | 119 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100773059| Gene Symbol: LOC100773059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509677.1 | XP_003509725.1 | 2 - 3 | 165 | 119 | |
Set Membership: |
|