Assay Details
| Gene Symbol: Ccl28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645453.1 | XP_007643643.1 | 1 - 2 | 51 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccl28| Gene Symbol: Ccl28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645453.1 | XP_007643643.1 | 1 - 2 | 51 | 64 | |
Set Membership: |
|