Assay Details
| Gene Symbol: Sar1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499885.2 | XP_003499933.1 | 2 - 3 | 182 | 90 | |
| GenBank mRNA | JP062053.1 | - | 2 - 3 | 179 | 90 | |
More Information
Set Membership: |
|

Sar1a
Genomic Map