Assay Details
| Gene Symbol: Kiaa0753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646884.1 | XP_007645074.1 | 18 - 19 | 3344 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kiaa0753| Gene Symbol: Kiaa0753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646884.1 | XP_007645074.1 | 18 - 19 | 3344 | 66 | |
Set Membership: |
|