Assay Details
| Gene Symbol: Rab13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501624.2 | XP_003501672.1 | 3 - 4 | 378 | 97 | |
| XM_007642286.1 | XP_007640476.1 | 3 - 4 | 485 | 97 | ||
| XM_007642287.1 | XP_007640477.1 | 3 - 4 | 435 | 97 | ||
| XM_007642288.1 | XP_007640478.1 | 3 - 4 | 321 | 97 | ||
| GenBank mRNA | JP039612.1 | - | 3 - 4 | 364 | 97 | |
More Information
Set Membership: |
|

Rab13
Genomic Map