Assay Details
| Gene Symbol: Il23a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506183.2 | XP_003506231.1 | 2 - 3 | 993 | 61 | |
| XM_007646671.1 | XP_007644861.1 | 3 - 4 | 883 | 61 | ||
More Information
Set Membership: |
|

Il23a
Genomic Map