Assay Details
| Gene Symbol: LOC100760504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647586.1 | XP_007645776.1 | 9 - 10 | 809 | 82 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100760504| Gene Symbol: LOC100760504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647586.1 | XP_007645776.1 | 9 - 10 | 809 | 82 | |