Assay Details
| Gene Symbol: Setd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644056.1 | XP_007642246.1 | 7 - 8 | 755 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Setd9| Gene Symbol: Setd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644056.1 | XP_007642246.1 | 7 - 8 | 755 | 63 | |
Set Membership: |
|