Assay Details
| Gene Symbol: Pak7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495251.1 | XP_003495299.1 | 6 - 7 | 1868 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pak7| Gene Symbol: Pak7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495251.1 | XP_003495299.1 | 6 - 7 | 1868 | 84 | |
Set Membership: |
|