Assay Details
| Gene Symbol: Josd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510202.2 | XP_003510250.1 | 2 - 3 | 318 | 84 | |
| XM_007650825.1 | XP_007649015.1 | 3 - 4 | 379 | 84 | ||
| XM_007650826.1 | XP_007649016.1 | 3 - 4 | 376 | 84 | ||
| XM_007650827.1 | XP_007649017.1 | 2 - 3 | 319 | 84 | ||
| XM_007650828.1 | XP_007649018.1 | 2 - 3 | 181 | 84 | ||
| GenBank mRNA | JP059902.1 | - | 2 - 3 | 289 | 84 | |
More Information
Set Membership: |
|

Josd2
Genomic Map