Assay Details
| Gene Symbol: Tmprss11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651100.1 | XP_007649290.1 | 6 - 7 | 713 | 96 | |
| XM_007651101.1 | XP_007649291.1 | 3 - 4 | 312 | 96 | ||
More Information
Set Membership: |
|

Tmprss11b
Genomic Map