Assay Details
| Gene Symbol: LOC100773612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504057.2 | XP_003504105.1 | 8 - 9 | 723 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100773612| Gene Symbol: LOC100773612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504057.2 | XP_003504105.1 | 8 - 9 | 723 | 61 | |
Set Membership: |
|