Assay Details
| Gene Symbol: Sult1c3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514253.2 | XP_003514301.1 | 6 - 7 | 802 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult1c3| Gene Symbol: Sult1c3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514253.2 | XP_003514301.1 | 6 - 7 | 802 | 71 | |
Set Membership: |
|