Assay Details
| Gene Symbol: Ube2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007624394.1 | XP_007622584.1 | 2 - 3 | 243 | 80 | |
| GenBank mRNA | JI868354.1 | - | 2 - 3 | 256 | 80 | |
| JP060122.1 | - | 2 - 3 | 256 | 80 | ||
More Information
Set Membership: |
|

Ube2e1
Genomic Map