Assay Details
| Gene Symbol: Smdt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508803.2 | XP_003508851.1 | 2 - 3 | 446 | 63 | |
| GenBank mRNA | JP050245.1 | - | 1 - 2 | 139 | 63 | |
More Information
Set Membership: |
|

Smdt1
Genomic Map