Assay Details
| Gene Symbol: LOC100750822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513759.2 | XP_003513807.1 | 10 - 11 | 1064 | 77 | |
| GenBank mRNA | JI882613.1 | - | 10 - 11 | 1046 | 77 | |
| JP045581.1 | - | 10 - 11 | 1046 | 77 | ||
More Information
Set Membership: |
|

LOC100750822
Genomic Map