Assay Details
| Gene Symbol: LOC100756805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654844.1 | XP_007653034.1 | 269 | 88 | ||
| GenBank mRNA | JI865404.1 | - | 269 | 88 | ||
More Information
Set Membership: |
|

LOC100756805
Genomic Map