Assay Details
| Gene Symbol: Rab21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640007.1 | XP_007638197.1 | 6 - 7 | 467 | 83 | |
| GenBank mRNA | JI893519.1 | - | 5 - 6 | 314 | 83 | |
More Information
Set Membership: |
|

Rab21
Genomic Map