Assay Details
| Gene Symbol: Igsf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502644.2 | XP_003502692.2 | 6 - 7 | 933 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igsf5| Gene Symbol: Igsf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502644.2 | XP_003502692.2 | 6 - 7 | 933 | 69 | |
Set Membership: |
|