Assay Details
| Gene Symbol: Aldoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510103.2 | XP_003510151.1 | 4 - 5 | 672 | 64 | |
| XM_003510105.2 | XP_003510153.1 | 4 - 5 | 442 | 64 | ||
| XM_007650737.1 | XP_007648927.1 | 4 - 5 | 537 | 64 | ||
| XM_007650738.1 | XP_007648928.1 | 4 - 5 | 464 | 64 | ||
| GenBank mRNA | JI884545.1 | - | 3 - 4 | 395 | 64 | |
| JP046793.1 | - | 3 - 4 | 395 | 64 | ||
More Information
Set Membership: |
|

Aldoa
Genomic Map