Assay Details
Gene Symbol: Plgrkt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511053.2 | XP_003511101.1 | 3 - 4 | 290 | 73 | |
XM_007651617.1 | XP_007649807.1 | 2 - 3 | 255 | 73 | ||
GenBank mRNA | JI888354.1 | - | 3 - 4 | 282 | 73 | |
JP056982.1 | - | 3 - 4 | 282 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
