Assay Details
| Gene Symbol: Hprt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643626.1 | XP_007641816.1 | 1 - 2 | 268 | 79 | |
| GenBank mRNA | JI892439.1 | - | 1 - 2 | 268 | 79 | |
More Information
Set Membership: |
|

Hprt1
Genomic Map