Assay Details
| Gene Symbol: Ube2t | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498853.2 | XP_003498901.1 | 6 - 7 | 664 | 67 | |
| GenBank mRNA | JI893611.1 | - | 6 - 7 | 632 | 67 | |
More Information
Set Membership: |
|

Ube2t
Genomic Map