Assay Details
| Gene Symbol: Gyg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508930.2 | XP_003508978.2 | 6 - 7 | 977 | 70 | |
| GenBank mRNA | JI868739.1 | - | 4 - 5 | 689 | 70 | |
| JP043392.1 | - | 4 - 5 | 688 | 70 | ||
More Information
Set Membership: |
|

Gyg1
Genomic Map