Assay Details
| Gene Symbol: Tmem106a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504618.2 | XP_003504666.1 | 4 - 5 | 584 | 61 | |
| XM_007645177.1 | XP_007643367.1 | 3 - 4 | 774 | 61 | ||
| XM_007645178.1 | XP_007643368.1 | 4 - 5 | 502 | 61 | ||
| GenBank mRNA | JI891757.1 | - | 4 - 5 | 548 | 61 | |
| JP048482.1 | - | 4 - 5 | 548 | 61 | ||
More Information
Set Membership: |
|

Tmem106a
Genomic Map