Assay Details
| Gene Symbol: Sppl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503513.2 | XP_003503561.1 | 7 - 8 | 758 | 66 | |
| GenBank mRNA | JI894284.1 | - | 7 - 8 | 769 | 66 | |
| JP049219.1 | - | 4 - 5 | 408 | 66 | ||
More Information
Set Membership: |
|

Sppl3
Genomic Map