Assay Details
| Gene Symbol: Aven | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503593.2 | XP_003503641.1 | 1 - 2 | 187 | 61 | |
| GenBank mRNA | JI892302.1 | - | 1 - 2 | 176 | 61 | |
More Information
Set Membership: |
|

Aven
Genomic Map