Assay Details
| Gene Symbol: Commd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507782.2 | XP_003507830.1 | 3 - 4 | 424 | 71 | |
| GenBank mRNA | JP041008.1 | - | 3 - 4 | 437 | 71 | |
More Information
Set Membership: |
|

Commd8
Genomic Map