Assay Details
| Gene Symbol: Noxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510560.2 | XP_003510608.1 | 7 - 8 | 941 | 58 | |
| XM_007651179.1 | XP_007649369.1 | 6 - 7 | 808 | 58 | ||
| GenBank mRNA | JI864363.1 | - | 2 - 3 | 70 | 58 | |
| JP051093.1 | - | 2 - 3 | 70 | 58 | ||
More Information
Set Membership: |
|

Noxa1
Genomic Map