Assay Details
| Gene Symbol: LOC100767953 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503218.2 | XP_003503266.2 | 1 - 2 | 137 | 68 | |
| XM_007643826.1 | XP_007642016.1 | 1 - 2 | 137 | 68 | ||
| XM_007643827.1 | XP_007642017.1 | 1 - 2 | 77 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_007643827.1 transcripts.
Set Membership: |
|

LOC100767953
Genomic Map