Assay Details
Gene Symbol: LOC100767953 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503218.2 | XP_003503266.2 | 2 - 3 | 202 | 72 | |
XM_007643826.1 | XP_007642016.1 | 2 - 3 | 202 | 72 | ||
XM_007643828.1 | XP_007642018.1 | 3 - 4 | 195 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
