Assay Details
| Gene Symbol: LOC100767953 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503218.2 | XP_003503266.2 | 3 - 4 | 295 | 68 | |
| XM_003503219.2 | XP_003503267.1 | 3 - 4 | 895 | 68 | ||
| XM_007643826.1 | XP_007642016.1 | 3 - 4 | 295 | 68 | ||
| XM_007643827.1 | XP_007642017.1 | 4 - 5 | 309 | 68 | ||
| XM_007643828.1 | XP_007642018.1 | 4 - 5 | 288 | 68 | ||
| XM_007643829.1 | XP_007642019.1 | 2 - 3 | 2754 | 68 | ||
| GenBank mRNA | JI878443.1 | - | 1 - 2 | 94 | 68 | |
| JP045029.1 | - | 1 - 2 | 94 | 68 | ||
More Information
Set Membership: |
|

LOC100767953
Genomic Map